Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-4-nitro-1H-indazole 97%

Supplier: AMBEED, INC
Warning

A243561-1G A243561-5G A243561-100MG A243561-10G A243561-250MG A243561-1G A243561-25G A243561-5G
103811-522EA 23 USD InStock
103811-522 103851-344 76976-522 76976-524 76976-526 77090-542 77090-544 77090-546
6-Bromo-4-nitro-1H-indazole 97%
6-Bromo-4-nitro-1H-indazole
Precaución: for Research Use Only


Formula: C₇H₄BrN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD07368216
CAS Number: 885518-46-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty