Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoimidazo[1,2-b]pyridazine 95%

Supplier: AMBEED, INC
Warning

A420569-1G A420569-5G A420569-100MG A420569-10G A420569-250MG A420569-1G
103827-388EA 18 USD InStock
103827-388 103838-436 76818-044 76818-046 76818-048 76842-060
6-Bromoimidazo[1,2-b]pyridazine 95%
6-Bromoimidazo[1,2-b]pyridazine
Precaución: for Research Use Only


Formula: C₆H₄BrN₃
MW: 198.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12198219
CAS Number: 1159977-65-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty