Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2,3-dihydro-1H-indole 98%

Supplier: AMBEED, INC
Warning

A221967-1G A221967-5G A221967-10G A221967-250MG A221967-1G A221967-5G
103818-956EA 16 USD InStock
103818-956 103849-832 77013-284 77013-286 77303-618 77303-620
6-Bromo-2,3-dihydro-1H-indole 98%
6-Bromo-2,3-dihydro-1H-indole
Precaución: for Research Use Only


Formula: C₈H₈BrN
Boiling Pt: 266.3 °C (760 mm Hg)
Melting Pt: 43…47 °C
Flash Pt: 114.869 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371638
CAS Number: 63839-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty