Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoisobenzofuran-1(3H)-one 97%

Supplier: AMBEED, INC
Warning

A337689-5G A337689-100G A337689-10G A337689-250MG A337689-1G A337689-25G A337689-5G
103825-752EA 45 USD InStock
103825-752 103836-724 76788-252 76788-254 76788-256 76839-736 76839-738 76839-740
6-Bromoisobenzofuran-1(3H)-one 97%
6-Bromoisobenzofuran-1(3H)-one
Precaución: for Research Use Only


Formula: C₈H₅BrO₂
Flash Pt: 173 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03428561
CAS Number: 19477-73-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty