Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1H-imidazo[4,5-c]pyridine 96%

Supplier: AMBEED, INC
Warning

A256224-1G A256224-100MG A256224-250MG
103812-252EA 251 USD InStock
103812-252 103852-116 76976-830 76976-832
6-Chloro-1H-imidazo[4,5-c]pyridine 96%
6-Chloro-1H-imidazo[4,5-c]pyridine
Precaución: for Research Use Only


Formula: C₆H₄ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD12022343
CAS Number: 2589-11-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty