Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloroindole 98%

Supplier: AMBEED, INC
Warning

A136917-1G A136917-100G A136917-10G A136917-1G A136917-25G A136917-5G
103802-374EA 5 USD InStock
103802-374 77272-296 77272-298 77298-874 77315-298 77315-300
6-Chloroindole 98%
6-Chloroindole
Precaución: for Research Use Only


Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 293 °C (760 mm Hg)
Melting Pt: 90
Storage Temperature: Ambient
MDL Number: MFCD00005681
CAS Number: 17422-33-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty