Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-1H-indole-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A170254-1G A170254-5G A170254-10G A170254-1G A170254-5G A170254-250MG A170254-25G
103806-466EA 7 USD InStock
103806-466 103844-734 76769-196 77081-838 77081-840 77877-115 77877-116
6-Chloro-1H-indole-3-carboxylic acid 98%
6-Chloro-1H-indole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₆ClNO₂
Flash Pt: 225.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07776607
CAS Number: 766557-02-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty