Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-6-chloro-2-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A216707-1G A216707-5G A216707-10G A216707-1G A216707-25G A216707-5G
103818-578EA 9 USD InStock
103818-578 103849-300 76999-808 77087-896 77087-898 77087-900
3-Amino-6-chloro-2-methylpyridine 98%
3-Amino-6-chloro-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₇ClN₂
Boiling Pt: 286 °C (760 mm Hg)
Flash Pt: 126.7 °C
Storage Temperature: Freezer
MDL Number: MFCD03095220
CAS Number: 164666-68-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty