Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-oxo-1,2-dihydropyridine-4-carboxylic acid 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A129892-1G A129892-1G A129892-250MG
103801-344EA 69 USD InStock
103801-344 77313-748 77683-152
6-Chloro-2-oxo-1,2-dihydropyridine-4-carboxylic acid 97%, Ambeed.Inc
6-Chloro-2-oxo-1,2-dihydropyridine-4-carboxylic acid
Precaución: For research use only.


Formula: C₆H₄ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07437951
CAS Number: 6313-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty