Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-(trifluoromethyl)picolinonitrile 95%

Supplier: AMBEED, INC
Warning

A204808-1G A204808-5G A204808-100MG A204808-250MG
103817-568EA 62 USD InStock
103817-568 103848-234 76825-680 76825-682
6-Chloro-3-(trifluoromethyl)picolinonitrile 95%
6-Chloro-3-(trifluoromethyl)picolinonitrile
Precaución: for Research Use Only


Formula: C₇H₂ClF₃N₂
Boiling Pt: 258.5±35.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD18261725
CAS Number: 401590-41-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty