Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-(trifluoromethyl)pyridine-2-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A430405-5G A430405-10G A430405-250MG A430405-25G A430405-1G A430405-5G
103827-508EA 99 USD InStock
103827-508 103838-572 76773-904 76773-906 76773-908 76842-258 77307-248
6-Chloro-3-(trifluoromethyl)pyridine-2-carboxylic acid 95%
6-Chloro-3-(trifluoromethyl)pyridine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₃ClF₃NO₂
MW: 225.5500 g/mol
Melting Pt: 303.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235206
CAS Number: 796090-24-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty