Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-3-methyl-1H-pyrazolo[4,3-c]pyridine 95%

Supplier: AMBEED, INC
Warning

A218212-1G A218212-5G A218212-100MG A218212-10G A218212-250MG A218212-1G A218212-5G
103818-700EA 66 USD InStock
103818-700 103849-428 76975-754 76975-756 76975-758 77088-090 77088-092
6-Chloro-3-methyl-1H-pyrazolo[4,3-c]pyridine 95%
6-Chloro-3-methyl-1H-pyrazolo[4,3-c]pyridine
Precaución: for Research Use Only


Formula: C₇H₆ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD16619899
CAS Number: 1092062-74-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty