Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-4-methoxypicolinic acid 95%

Supplier: AMBEED, INC
Warning

A252213-1G A252213-5G A252213-100MG A252213-250MG A252213-1G
103812-034EA 127 USD InStock
103812-034 103851-876 76815-684 76815-686 77091-286
6-Chloro-4-methoxypicolinic acid 95%
6-Chloro-4-methoxypicolinic acid
Precaución: for Research Use Only


Formula: C₇H₆ClNO₃
Boiling Pt: 322.6 °C (760 mm Hg)
Flash Pt: 148.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03844441
CAS Number: 88912-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty