Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-5-nitropyridin-2-amine 98%

Supplier: AMBEED, INC
Warning

A129272-1G A129272-100MG A129272-250MG
103801-252EA 129 USD InStock
103801-252 76870-646 76870-648
6-Chloro-5-nitropyridin-2-amine 98%
6-Chloro-5-nitropyridin-2-amine
Precaución: for Research Use Only


Formula: C₅H₄ClN₃O₂
MW: 173.56 g/mol
Boiling Pt: 393.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01692500
CAS Number: 84487-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty