Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chlorohexanoic acid 95%

Supplier: AMBEED, INC
Danger

A204292-1G A204292-10G A204292-1G A204292-25G A204292-5G
103817-540EA 11 USD InStock
103817-540 103848-202 76770-276 77086-362 77086-364 77086-366
6-Chlorohexanoic acid 95%
6-Chlorohexanoic acid
Precaución: for Research Use Only


Formula: C₆H₁₁ClO₂
MW: 150.60 g/mol
Boiling Pt: 254.4 °C (760 mm Hg)
Melting Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061103
CAS Number: 4224-62-8
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty