Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine 98%

Supplier: AMBEED, INC
Warning

A148484-1G A148484-5G A148484-25G A148484-10G
103803-932EA 16 USD InStock
103803-932 103842-044 77299-660 77881-160
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine 98%
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine
Precaución: for Research Use Only


Formula: C₆H₈ClN₃
Boiling Pt: 248.7±35.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD12024889
CAS Number: 61655-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty