Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloropurine ribonucleoside 97%

Supplier: AMBEED, INC
Warning

A516903-5G A516903-1G A516903-25G A516903-10G A516903-5G
103828-466EA 9 USD InStock
103828-466 103839-572 76843-560 76843-562 76991-426 77307-686
6-Chloropurine ribonucleoside 97%
6-Chloropurine ribonucleoside
Precaución: for Research Use Only


Formula: C₁₀H₁₁ClN₄O₄
MW: 286.672 g/mol
Boiling Pt: 614.8 °C (760 mm Hg)
Melting Pt: 161…163 °C
Flash Pt: 325.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00005738
CAS Number: 2004-06-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty