Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloropyridine-3-sulfonamide 95%

Supplier: AMBEED, INC
Warning

A192628-1G A192628-5G A192628-100MG A192628-250MG A192628-5G
103816-468EA 36 USD InStock
103816-468 103847-078 76974-912 76974-914 77302-314
6-Chloropyridine-3-sulfonamide 95%
6-Chloropyridine-3-sulfonamide
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O₂S
Boiling Pt: 383.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00661410
CAS Number: 40741-46-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty