Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-4-quinolinol 97%

Supplier: AMBEED, INC
Warning

A241827-1G A241827-5G A241827-1G A241827-25G A241827-100G A241827-10G
103811-430EA 7 USD InStock
103811-430 103851-236 77090-398 77090-400 77261-806 77261-808
6-Chloro-4-quinolinol 97%
6-Chloro-4-quinolinol
Precaución: for Research Use Only


Formula: C₉H₆ClNO
MW: 179.60 g/mol
Boiling Pt: 306.9 °C (760 mm Hg)
Flash Pt: 139.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00024011
CAS Number: 23432-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty