Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoro-1-tetralone 97%

Supplier: AMBEED, INC
Warning

A124542-1G A124542-250MG A124542-1G A124542-5G
103800-546EA 21 USD InStock
103800-546 76810-746 77312-682 77312-684
6-Fluoro-1-tetralone 97%
6-Fluoro-1-tetralone
Precaución: For research use only.


Formula: C₁₀H₉FO
Boiling Pt: 269.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09031370
CAS Number: 703-67-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty