Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluorobenzo[b]thiophene 95%

Supplier: AMBEED, INC
Warning

A584862-5G A584862-1G A584862-100MG A584862-250MG
103828-880EA 149 USD InStock
103828-880 103840-026 76844-160 76980-406 76980-408
6-Fluorobenzo[b]thiophene 95%
6-Fluorobenzo[b]thiophene
Precaución: For research use only


Formula: C₈H₅FS
Storage Temperature: Refrigerator
MDL Number: MFCD18207210
CAS Number: 205055-10-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty