Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Fluoroindole-3-carboxaldehyde 98%

Supplier: AMBEED, INC
Warning

A106458-1G A106458-5G A106458-100G A106458-10G A106458-250MG A106458-5G A106458-1G
103797-036EA 20 USD InStock
103797-036 103822-790 76687-262 76687-264 76687-266 77283-480 77309-644
6-Fluoroindole-3-carboxaldehyde 98%
6-Fluoroindole-3-carboxaldehyde
Precaución: for Research Use Only


Formula: C₉H₆FNO
MW: 163.15 g/mol
Boiling Pt: 342.4 °C (760 mm Hg)
Melting Pt: 174…178 °C
Flash Pt: 160.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00069703
CAS Number: 2795-41-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty