Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Mercaptopurine monohydrate 98%

Supplier: AMBEED, INC
Warning

A189537-5G A189537-100MG A189537-10G A189537-25G A189537-100G
103846-790EA 10 USD InStock
103846-790 76813-866 76813-868 77302-182 77671-690
6-Mercaptopurine monohydrate 98%
6-Mercaptopurine monohydrate
Precaución: For research use only.


Formula: C₅H₄N₄S·H₂O
Boiling Pt: 490.6 °C (760 mm Hg)
Melting Pt: 313 °C
Flash Pt: 250.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03854445
CAS Number: 6112-76-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty