Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-2-(trifluoromethyl)-1H-benzo[d]imidazole 97%

Supplier: AMBEED, INC
Warning

A121472-1G A121472-100MG A121472-10G A121472-1G A121472-5G
103800-080EA 57 USD InStock
103800-080 76766-512 76766-514 77297-834 77297-836
6-Methoxy-2-(trifluoromethyl)-1H-benzo[d]imidazole 97%
6-Methoxy-2-(trifluoromethyl)-1H-benzo[d]imidazole
Precaución: for Research Use Only


Formula: C₉H₇F₃N₂O
Boiling Pt: 307 °C (760 mm Hg)
Flash Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09264131
CAS Number: 3671-65-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty