Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxy-3-methylisoquinoline 98%

Supplier: AMBEED, INC
Warning

A490705-1G A490705-5G A490705-100MG A490705-250MG A490705-1G
103828-216EA 64 USD InStock
103828-216 103839-288 77017-128 77017-130 77307-584
6-Methoxy-3-methylisoquinoline 98%
6-Methoxy-3-methylisoquinoline
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO
MW: 173.211 g/mol
Boiling Pt: 301.1±22.0 °C (760 mm Hg)
Flash Pt: 110.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01846013
CAS Number: 14446-31-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty