Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxyimidazo[1,2-a]pyridine-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A189608-1G A189608-5G A189608-100MG A189608-250MG
103816-210EA 133 USD InStock
103816-210 103846-804 77164-192 77164-194
6-Methoxyimidazo[1,2-a]pyridine-2-carboxylic acid 97%
6-Methoxyimidazo[1,2-a]pyridine-2-carboxylic acid
Precaución: Sealed in dry, at 2…8°C


Formula: C₉H₈N₂O₃
MW: 192.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12401188
CAS Number: 1159832-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty