Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxyindole 98%

Supplier: AMBEED, INC
Warning

A129855-1G A129855-100G A129855-10G A129855-1G A129855-25G A129855-5G
103801-342EA 15 USD InStock
103801-342 77271-956 77271-958 77313-742 77313-744 77313-746
6-Methoxyindole 98%
6-Methoxyindole
Precaución: for Research Use Only


Formula: C₉H₉NO
Boiling Pt: 297.8 °C (760 mm Hg)
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00022780
CAS Number: 3189-13-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty