Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylbenzo[b]thiophene 98%

Supplier: AMBEED, INC
Warning

A298899-1G A298899-5G A298899-100MG A298899-250MG A298899-1G
103814-200EA 198 USD InStock
103814-200 103835-348 76772-590 76772-592 76838-052
6-Methylbenzo[b]thiophene 98%
6-Methylbenzo[b]thiophene
Precaución: for Research Use Only


Formula: C₉H₈S
MW: 148.23 g/mol
Boiling Pt: 243 °C (760 mm Hg)
Melting Pt: 42.0…42.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02683997
CAS Number: 16587-47-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty