Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methylquinoxaline 95%

Supplier: AMBEED, INC
Warning

A173049-1G A173049-5G A173049-100MG A173049-250MG A173049-1G A173049-5G
103806-784EA 19 USD InStock
103806-784 103845-078 76813-272 76813-274 76813-276 77301-240 77301-242
6-Methylquinoxaline 95%
6-Methylquinoxaline
Precaución: for Research Use Only


Formula: C₉H₈N₂
Boiling Pt: 257.2 °C (760 mm Hg)
Flash Pt: 108.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00041001
CAS Number: 6344-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty