Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxypyridazine-3-carboxaldehyde ≥95%

Supplier: AMBEED, INC
Warning

A220180-1G A220180-100MG A220180-250MG
103818-840EA 320 USD InStock
103818-840 103849-722 77872-115 77872-116
6-Hydroxypyridazine-3-carboxaldehyde ≥95%
6-Hydroxypyridazine-3-carboxaldehyde
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₅H₄N₂O₂
Boiling Pt: 398.8 °C (760 mm Hg)
Flash Pt: 195 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09998728
CAS Number: 933734-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty