Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Oxo-1,6-dihydropyridine-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A131750-1G A131750-100G A131750-10G A131750-1G A131750-25G A131750-5G
103801-626EA 9 USD InStock
103801-626 77272-036 77272-038 77314-142 77314-144 77314-146
6-Oxo-1,6-dihydropyridine-2-carboxylic acid 97%
6-Oxo-1,6-dihydropyridine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₅NO₃
Boiling Pt: 436 °C (760 mm Hg)
Melting Pt: 270 °C
Flash Pt: 217.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03548246
CAS Number: 19621-92-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty