Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid 97%

Supplier: AMBEED, INC
Warning

A261926-5G A261926-100G A261926-10G A261926-250MG A261926-1G A261926-25G A261926-5G
103812-546EA 18 USD InStock
103812-546 103852-438 76722-820 76722-822 76722-824 77092-054 77092-056 77092-058
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid 97%
N-(tert-Butoxycarbonyl)-7-aminoheptanoic acid
Precaución: for Research Use Only


Formula: C₁₂H₂₃NO₄
Boiling Pt: 393.1 °C (760 mm Hg)
Melting Pt: 56…60 °C
Flash Pt: 191.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063356
CAS Number: 60142-89-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty