Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-(Benzyloxy)quinolin-4-ol 95%

Supplier: AMBEED, INC
Warning

A121418-1G A121418-10G A121418-250MG A121418-1G A121418-5G
103800-070EA 42 USD InStock
103800-070 76971-542 76971-544 77312-190 77312-192
7-(Benzyloxy)quinolin-4-ol 95%
7-(Benzyloxy)quinolin-4-ol
Precaución: for Research Use Only


Formula: C₁₆H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD06659963
CAS Number: 749922-34-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty