Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Aminocephalosporanic acid 98%

Supplier: AMBEED, INC
Warning

A183629-5G A183629-100G A183629-500G A183629-25G A183629-1000G A183629-10G
103846-214EA 6 USD InStock
103846-214 76669-760 76669-762 76669-764 77083-822 77301-838 77625-870 77876-775
7-Aminocephalosporanic acid 98%
7-Aminocephalosporanic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₂O₅S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005177
CAS Number: 957-68-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty