Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Aminoenanthic acid 97%

Supplier: AMBEED, INC
Warning

A251386-1G A251386-100G A251386-10G A251386-500G A251386-1G A251386-5G A251386-25G
103811-980EA 10 USD InStock
103811-980 103851-822 76771-648 76771-650 76771-652 77091-200 77091-202 77304-556
7-Aminoenanthic acid 97%
7-Aminoenanthic acid


Formula: C₇H₁₅NO₂
Boiling Pt: 270.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00008242
CAS Number: 929-17-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty