Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Benzyl-4-chloro-7H-pyrrolo[2,3-d] pyrimidine 95%

Supplier: AMBEED, INC
Warning

A317351-5G A317351-1G A317351-250MG
103825-130EA 19 USD InStock
103825-130 103836-062 76838-910 77154-470
7-Benzyl-4-chloro-7H-pyrrolo[2,3-d] pyrimidine 95%
7-Benzyl-4-chloro-7H-pyrrolo[2,3-d] pyrimidine
Precaución: for Research Use Only


Formula: C₁₃H₁₀ClN₃
MW: 243.69 g/mol
Boiling Pt: 425.7±35.0 °C (760 mm Hg)
Melting Pt: 68…71 °C
Flash Pt: 211.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09953011
CAS Number: 16019-34-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty