Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-2-chloroquinoxaline 97%

Supplier: AMBEED, INC
Warning

A126981-1G A126981-100MG A126981-10G A126981-250MG A126981-1G A126981-5G
103800-892EA 9 USD InStock
103800-892 76688-468 76688-470 76688-472 77313-138 77313-140
7-Bromo-2-chloroquinoxaline 97%
7-Bromo-2-chloroquinoxaline
Precaución: for Research Use Only


Formula: C₈H₄BrClN₂
Melting Pt: 146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02955309
CAS Number: 89891-65-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty