Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-5-chloro-1H-indole 95%

Supplier: AMBEED, INC
Warning

A160543-1G A160543-5G A160543-100MG A160543-10G A160543-250MG A160543-5G A160543-1G
103805-392EA 43 USD InStock
103805-392 103843-624 76783-804 76783-806 76783-808 77300-474 77319-568
7-Bromo-5-chloro-1H-indole 95%
7-Bromo-5-chloro-1H-indole
Precaución: for Research Use Only


Formula: C₈H₅BrClN
MW: 230.48 g/mol
Boiling Pt: 348.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09880061
CAS Number: 292636-08-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty