Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromo-5H-pyrrolo[3,2-d]pyrimidine 95%

Supplier: AMBEED, INC
Warning

A327738-1G A327738-100MG A327738-250MG
103825-476EA 420 USD InStock
103825-476 103836-426 77015-366 77015-368
7-Bromo-5H-pyrrolo[3,2-d]pyrimidine 95%
7-Bromo-5H-pyrrolo[3,2-d]pyrimidine
Precaución: for Research Use Only


Formula: C₆H₄BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD19687341
CAS Number: 1378816-68-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty