Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Bromoquinazolin-4(3H)-one 98%

Supplier: AMBEED, INC
Warning

A235892-5G A235892-100G A235892-10G A235892-1G A235892-25G A235892-5G
103811-042EA 5 USD InStock
103811-042 103850-812 76692-164 76692-166 77089-830 77089-832 77089-834
7-Bromoquinazolin-4(3H)-one 98%
7-Bromoquinazolin-4(3H)-one
Precaución: for Research Use Only


Formula: C₈H₅BrN₂O
Boiling Pt: 428.7 °C (760 mm Hg)
Flash Pt: 213.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07838323
CAS Number: 194851-16-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty