Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-1,2,3,4-tetrahydroisoquinoline 97%

Supplier: AMBEED, INC
Warning

A433173-1G A433173-5G A433173-100MG A433173-250MG A433173-1G
103827-544EA 174 USD InStock
103827-544 103838-610 76648-962 76648-964 77307-262
7-Chloro-1,2,3,4-tetrahydroisoquinoline 97%
7-Chloro-1,2,3,4-tetrahydroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₁₀ClN
Boiling Pt: 267.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD05861543
CAS Number: 82771-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty