Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A129712-1000G A129712-100G A129712-10G A129712-500G A129712-25G A129712-5G
76718-902EA 140 USD InStock
76718-902 76718-904 76718-906 76718-908 77313-702 77313-704
7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%
7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₉ClFNO₃
Melting Pt: 242…245 °C
Flash Pt: 236.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00792458
CAS Number: 86393-33-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty