Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-1H-pyrrolo[3,2-b]pyridine 95%

Supplier: AMBEED, INC
Warning

A111545-1G A111545-5G A111545-250MG A111545-25G A111545-1G A111545-5G
103797-856EA 32 USD InStock
103797-856 103823-674 76687-616 76687-618 77310-480 77310-482
7-Chloro-1H-pyrrolo[3,2-b]pyridine 95%
7-Chloro-1H-pyrrolo[3,2-b]pyridine
Precaución: for Research Use Only


Formula: C₇H₅ClN₂
Boiling Pt: 290.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08234945
CAS Number: 357263-48-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty