Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Chloro-8-methoxyquinoxalin-2-ol 95%

Supplier: AMBEED, INC
Warning

A318973-1G A318973-100MG A318973-250MG A318973-1G
103825-188EA 361 USD InStock
103825-188 77015-242 77015-244 77306-194
7-Chloro-8-methoxyquinoxalin-2-ol 95%
7-Chloro-8-methoxyquinoxalin-2-ol
Precaución: for Research Use Only


Formula: C₉H₇ClN₂O₂
Melting Pt: 170…172 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11846479
CAS Number: 347162-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty