Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Fluoro-1-tetralone 97%

Supplier: AMBEED, INC
Warning

A306552-1G A306552-1G A306552-25G A306552-5G A306552-100G A306552-10G A306552-250MG
103814-476EA 12 USD InStock
103814-476 103835-642 76838-360 76838-362 76838-364 77002-588 77002-590 77002-592
7-Fluoro-1-tetralone 97%
7-Fluoro-1-tetralone
Precaución: for Research Use Only


Formula: C₁₀H₉FO
MW: 164.18 g/mol
Boiling Pt: 258.1 °C (760 mm Hg)
Melting Pt: 61…66 °C
Flash Pt: 99.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07375433
CAS Number: 2840-44-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty