Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Fluoro-6-nitroquinazolin-4(3H)-one 98%

Supplier: AMBEED, INC
Warning

A134329-1G A134329-100G A134329-10G A134329-250MG A134329-500G A134329-1G A134329-25G A134329-5G
103802-004EA 9 USD InStock
103802-004 76996-574 76996-576 76996-578 76996-580 77314-742 77314-744 77314-746
7-Fluoro-6-nitroquinazolin-4(3H)-one 98%
7-Fluoro-6-nitroquinazolin-4(3H)-one
Precaución: for Research Use Only


Formula: C₈H₄FN₃O₃
Boiling Pt: 407.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08690890
CAS Number: 162012-69-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty