Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Hydroxychroman-4-one 97%

Supplier: AMBEED, INC
Warning

A135329-1G A135329-100MG A135329-250MG A135329-1G A135329-5G
103802-132EA 81 USD InStock
103802-132 77285-480 77285-482 77314-946 77747-966
7-Hydroxychroman-4-one 97%
7-Hydroxychroman-4-one
Precaución: for Research Use Only


Formula: C₉H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD08669475
CAS Number: 76240-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty