Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Methyl-1H-indole-3-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A175020-1G A175020-5G A175020-10G A175020-1G A175020-25G A175020-250MG
103806-994EA 14 USD InStock
103806-994 103845-294 76690-300 77301-360 77301-362 77625-828
7-Methyl-1H-indole-3-carboxylic acid 97%
7-Methyl-1H-indole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₉NO₂
MW: 175.1800 g/mol
Melting Pt: 238…241 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06203634
CAS Number: 30448-16-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty