Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Bromoimidazo[1,2-a]pyridine 95%

Supplier: AMBEED, INC
Warning

A216086-1G A216086-5G A216086-1G A216086-5G A216086-100MG A216086-10G A216086-250MG
103818-534EA 27 USD InStock
103818-534 103849-246 77087-816 77087-818 77165-094 77165-096 77165-098
8-Bromoimidazo[1,2-a]pyridine 95%
8-Bromoimidazo[1,2-a]pyridine
Precaución: for Research Use Only


Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Melting Pt: 70…75 °C
Storage Temperature: Ambient
MDL Number: MFCD08706060
CAS Number: 850349-02-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty