Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Bromoquinoline-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A336226-5G A336226-1G A336226-250MG
103825-712EA 94 USD InStock
103825-712 103836-688 76839-694 76978-358
8-Bromoquinoline-5-carboxylic acid 97%
8-Bromoquinoline-5-carboxylic acid
Precaución: For research use only


Formula: C₁₀H₆BrNO₂
Boiling Pt: 425.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD12963555
CAS Number: 204782-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty